3,5-Dimethyl-1H-pyrazol-4-boronsäurepinakolester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 3,5-Dimethylpyrazole-4-boronic acid pinacol ester

OR2255-1G OR2255-5G OR2255-25G
APOSOR2255-1GEA 94.4 EUR
APOSOR2255-1G APOSOR2255-5G APOSOR2255-25G
3,5-Dimethyl-1H-pyrazol-4-boronsäurepinakolester 97%
3,5-Dimethyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₁H₁₉BN₂O₂
Molecular Weight: 222.0918 g/mol
Schmelzpunkt: 163…168 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05663852
CAS-Nummer: 857530-80-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR