(±)-1-Boc-3-hydroxypyrrolidin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 3-hydroxypyrrolidine-1-carboxylate, 3-Hydroxypyrrolidine, N-BOC protected
OR2256-25G
OR2256-5G
APOSOR2256-25GEA
37.7
EUR
APOSOR2256-25G
APOSOR2256-5G
(±)-1-Boc-3-hydroxypyrrolidin
(±)-1-Boc-3-hydroxypyrrolidin
Formel:
C₉H₁₇NO₃ Molecular Weight: 187,24 g/mol Schmelzpunkt: 57…63 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04038535 CAS-Nummer: 103057-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...