(4-Ethoxyphenyl)urea
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-Ethoxyphenyl)urea
OR22579-1G
OR22579-5G
OR22579-25G
APOSOR22579-1GEA
58
EUR
APOSOR22579-1G
APOSOR22579-5G
APOSOR22579-25G
(4-Ethoxyphenyl)urea
(4-Ethoxyphenyl)urea
Formel:
C₉H₁₂N₂O₂ Molecular Weight: 180,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025431 CAS-Nummer: 150-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...