N-(4-Aminophenyl)-N-methylacetamide

Lieferant: Apollo Scientific
Achtung

OR22594-250MG OR22594-100MG
APOSOR22594-250MGEA 21.9 EUR
APOSOR22594-250MG APOSOR22594-100MG
N-(4-Aminophenyl)-N-methylacetamide
N-(4-Aminophenyl)-N-methylacetamide
Formel: C₉H₁₂N₂O
Molecular Weight: 164.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007859
CAS-Nummer: 119-63-1
EINECS: 204-339-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR