3-Amino-2-brompyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Brom-3-pyridylamin, 2-Brompyridin-3-amin
OR2264-25G
OR2264-5G
OR2264-100G
APOSOR2264-25GEA
55.1
EUR
APOSOR2264-25G
APOSOR2264-5G
APOSOR2264-100G
3-Amino-2-brompyridin 98%
3-Amino-2-brompyridin
Formel:
C₅H₅BrN₂ Molecular Weight: 173,01 g/mol Schmelzpunkt: 76…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234064 CAS-Nummer: 39856-58-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...