2-(4-Pyrimidyl)malondialdehyde 95+%

Lieferant: Apollo Scientific
Gefahr

OR22712-5G OR22712-1G
APOSOR22712-5GEA 580 EUR
APOSOR22712-5G APOSOR22712-1G
2-(4-Pyrimidyl)malondialdehyde 95+%
2-(4-Pyrimidyl)malondialdehyde
Formel: C₇H₆N₂O₂
Molecular Weight: 150,137 g/mol
Schmelzpunkt: 227…232 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 28648-78-4
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR