3-Acetyl-2,5-dichlorthiophen

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2,5-Dichlor-3-thienyl)-1-ethanon

OR22717-25G OR22717-100G
APOSOR22717-25GEA 23.2 EUR
APOSOR22717-25G APOSOR22717-100G
3-Acetyl-2,5-dichlorthiophen
3-Acetyl-2,5-dichlorthiophen
Formel: C₆H₄Cl₂OS
Molecular Weight: 195.07 g/mol
Schmelzpunkt: 37…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014522
CAS-Nummer: 36157-40-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR