1-Chloro-4-methylpentane

Lieferant: Apollo Scientific

OR22928-1G OR22928-250MG OR22928-500MG OR22928-2.5G
APOSOR22928-1GEA 328 EUR
APOSOR22928-1G APOSOR22928-250MG APOSOR22928-500MG APOSOR22928-2.5G
1-Chloro-4-methylpentane
1-Chloro-4-methylpentane
Formel: C₆H₁₃Cl
Molecular Weight: 120.62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00045304
CAS-Nummer: 62016-94-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR