[5-Methyl-3-phenyl-4-isoxazolyl]methanol

Lieferant: Apollo Scientific
Achtung

OR22984-1G OR22984-250MG
APOSOR22984-1GEA 132 EUR
APOSOR22984-1G APOSOR22984-250MG
[5-Methyl-3-phenyl-4-isoxazolyl]methanol
[5-Methyl-3-phenyl-4-isoxazolyl]methanol
Formel: C₁₁H₁₁NO₂
Schmelzpunkt: 81…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02677724
CAS-Nummer: 18718-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR