5-Hydroxymethyl-2,3-dihydrobenzo[b]furan

Lieferant: Apollo Scientific
Achtung

Synonyme: (2,3-Dihydrobenzofuran-5-yl)methanol

OR22994-250MG OR22994-100MG
APOSOR22994-250MGEA 30.6 EUR
APOSOR22994-250MG APOSOR22994-100MG
5-Hydroxymethyl-2,3-dihydrobenzo[b]furan
5-Hydroxymethyl-2,3-dihydrobenzo[b]furan
Formel: C₉H₁₀O₂
Molecular Weight: 150,18 g/mol
Siedepunkt: 152…155 °C/8mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD02169755
CAS-Nummer: 103262-35-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR