4-(1H-Pyrazol-3-yl)benzonitril

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(4-Cyanophenyl)-1H-pyrazole

OR23-1G OR23-5G
APOSOR23-1GEA 72.5 EUR
APOSOR23-1G APOSOR23-5G
4-(1H-Pyrazol-3-yl)benzonitril
4-(1H-Pyrazol-3-yl)benzonitril
Formel: C₁₀H₇N₃
Molecular Weight: 169,19 g/mol
Schmelzpunkt: 140…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01569438
CAS-Nummer: 474706-35-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR