3-(Dimethylamino)benzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-(Dimethylamino)phenyl)methanol, m-(Dimethylamino)benzyl alcohol, Benzenemethanol,3-(dimethylamino)-, Benzyl alcohol, m-(dimethylamino)-

OR2300-5G OR2300-25G
APOSOR2300-5GEA 87 EUR
APOSOR2300-5G APOSOR2300-25G
3-(Dimethylamino)benzyl alcohol 95%
3-(Dimethylamino)benzyl alcohol
Formel: C₉H₁₃NO
Molecular Weight: 151.209 g/mol
Siedepunkt: 282…283 °C
Dichte: 1.056 g/cm³ (25 °C)
Flammpunkt: 110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017342
CAS-Nummer: 23501-93-1
EINECS: 245-700-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR