6-Isocyanato-2,3-dihydro-1,4-benzodioxine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydro-1,4-benzodioxin-6-yl isocyanate

OR23007-1G OR23007-5G
APOSOR23007-1GEA 161 EUR
APOSOR23007-1G APOSOR23007-5G
6-Isocyanato-2,3-dihydro-1,4-benzodioxine 95%
6-Isocyanato-2,3-dihydro-1,4-benzodioxine

  • May form small amount of sediment.

Formel: C₉H₇NO₃
Molecular Weight: 177.16 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD02677705
CAS-Nummer: 100275-94-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR