2,3-Dihydro-1,4-benzodioxin-6-ylmethanol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydro-6-(hydroxymethyl)-1,4-benzodioxine, (2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)methanol

OR23009-1G OR23009-250MG
APOSOR23009-1GEA 89.9 EUR
APOSOR23009-1G APOSOR23009-250MG
2,3-Dihydro-1,4-benzodioxin-6-ylmethanol
2,3-Dihydro-1,4-benzodioxin-6-ylmethanol
Formel: C₉H₁₀O₃
Molecular Weight: 166.18 g/mol
Siedepunkt: 129 °C/0.2mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD01317581
CAS-Nummer: 39270-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR