1,4-Benzodioxan-6-boronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydro-1,4-benzodioxine-6-boronic acid

OR23011-1G OR23011-5G
APOSOR23011-1GEA 21.9 EUR
APOSOR23011-1G APOSOR23011-5G
1,4-Benzodioxan-6-boronsäure
1,4-Benzodioxan-6-boronsäure
Formel: C₈H₉BO₄
Molecular Weight: 179,97 g/mol
Schmelzpunkt: 116…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01009696
CAS-Nummer: 164014-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR