5-(Pyridin-2-yl)thiophene-2-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR23033-1G
OR23033-250MG
OR23033-5G
APOSOR23033-1GEA
133
EUR
APOSOR23033-1G
APOSOR23033-250MG
APOSOR23033-5G
5-(Pyridin-2-yl)thiophene-2-carbaldehyde
5-(Pyridin-2-yl)thiophene-2-carbaldehyde
Formel:
C₁₀H₇NOS Molecular Weight: 189,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00114999 CAS-Nummer: 132706-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...