3-Amino-4-brombenzoesäure 98%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-4-bromobenzoic Acid

OR2304-25G
APOSOR2304-25GEA 89.9 EUR
APOSOR2304-25G
3-Amino-4-brombenzoesäure 98%
3-Amino-4-bromobenzoic acid
Formel: C₇H₆BrNO₂
Molecular Weight: 216,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00579106
CAS-Nummer: 2840-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR