3-Bromo-1-(phenylsulphonyl)indole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-1-(phenylsulphonyl)-1H-indole
OR23046-1G
OR23046-5G
OR23046-25G
APOSOR23046-1GEA
21.9
EUR
APOSOR23046-1G
APOSOR23046-5G
APOSOR23046-25G
3-Bromo-1-(phenylsulphonyl)indole
3-Bromo-1-(phenylsulphonyl)indole
Formel:
C₁₄H₁₀BrNO₂S Molecular Weight: 336.203 g/mol Schmelzpunkt: 122…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02681986 CAS-Nummer: 99655-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...