Isochinolin-1-ylmethanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(Hydroxymethyl)isoquinoline
OR23079-1G
OR23079-250MG
APOSOR23079-1GEA
562
EUR
APOSOR23079-1G
APOSOR23079-250MG
Isochinolin-1-ylmethanol
Isochinolin-1-ylmethanol
Formel:
C₁₀H₉NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD03086097 CAS-Nummer: 27311-63-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...