(2,4-Dimethyl-1,3-thiazol-5-yl)methanol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Dimethyl-5-(hydroxymethyl)-1,3-thiazole

OR23083-1G
APOSOR23083-1GEA 75.4 EUR
APOSOR23083-1G
(2,4-Dimethyl-1,3-thiazol-5-yl)methanol
(2,4-Dimethyl-1,3-thiazol-5-yl)methanol
Formel: C₆H₉NOS
Molecular Weight: 143,21 g/mol
Schmelzpunkt: 56…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03086098
CAS-Nummer: 50382-32-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR