Benzothiazole-2-carbonyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

OR23087-1G OR23087-5G
APOSOR23087-1GEA 109 EUR
APOSOR23087-1G APOSOR23087-5G
Benzothiazole-2-carbonyl chloride 97%
Benzothiazole-2-carbonyl chloride
Formel: C₈H₄ClNOS
Molecular Weight: 197.64 g/mol
Schmelzpunkt: 124…130 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03659697
CAS-Nummer: 67748-61-2
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR