5-Benzofurancarbonsäure 95%

Lieferant: Apollo Scientific
Achtung

OR23098-1G OR23098-250MG
APOSOR23098-1GEA 33.5 EUR
APOSOR23098-1G APOSOR23098-250MG
5-Benzofurancarbonsäure 95%
Benzofuran-5-carboxylic acid
Formel: C₉H₆O₃
Molecular Weight: 162,1400 g/mol
Schmelzpunkt: 189…190 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01006742
CAS-Nummer: 90721-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR