2,1,3-Benzothiadiazol-5-carbaldehyd

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzo[c][1,2,5]thiadiazole-5-carbaldehyde

OR23127-1G OR23127-5G
APOSOR23127-1GEA 145 EUR
APOSOR23127-1G APOSOR23127-5G
2,1,3-Benzothiadiazol-5-carbaldehyd
2,1,3-Benzothiadiazol-5-carbaldehyd
Formel: C₇H₄N₂OS
Molecular Weight: 164,1800 g/mol
Schmelzpunkt: 89…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00457949
CAS-Nummer: 71605-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR