2,1,3-Benzothiadiazol-5-ylisothiocyanat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Isothiocyanato-2,1,3-benzothiadiazole
OR23128-1G
OR23128-250MG
APOSOR23128-1GEA
418
EUR
APOSOR23128-1G
APOSOR23128-250MG
2,1,3-Benzothiadiazol-5-ylisothiocyanat
2,1,3-Benzothiadiazol-5-ylisothiocyanat
Formel:
C₇H₃N₃S₂ Schmelzpunkt: 96…97 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02681895 CAS-Nummer: 337508-62-0 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...