5-(Brommethyl)-2,1,3-benzothiadiazol
Lieferant: Apollo Scientific
|
Gefahr
|
OR23129-1G
OR23129-250MG
APOSOR23129-1GEA
145
EUR
APOSOR23129-1G
APOSOR23129-250MG
5-(Brommethyl)-2,1,3-benzothiadiazol
5-(Bromomethyl)-2,1,3-benzothiadiazole
Formel:
C₇H₅BrN₂S Schmelzpunkt: 87…91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00544836 CAS-Nummer: 65858-50-6 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...