5-Brom-2,1,3-benzothiadiazol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromobenzo[c][1,2,5]thiadiazole
OR23131-1G
OR23131-250MG
OR23131-5G
APOSOR23131-1GEA
34.8
EUR
APOSOR23131-1G
APOSOR23131-250MG
APOSOR23131-5G
5-Brom-2,1,3-benzothiadiazol
5-Bromo-2,1,3-benzothiadiazole
Formel:
C₆H₃BrN₂S Molecular Weight: 215,0700 g/mol Schmelzpunkt: 59…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00460091 CAS-Nummer: 1753-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...