5-Bromobenzo[c][1,2,5]oxadiazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-2,1,3-benzoxadiazole
OR23136-1G
OR23136-5G
APOSOR23136-1GEA
29
EUR
APOSOR23136-1G
APOSOR23136-5G
5-Bromobenzo[c][1,2,5]oxadiazole
5-Bromobenzo[c][1,2,5]oxadiazole
Formel:
C₆H₃BrN₂O Molecular Weight: 199.01 g/mol Schmelzpunkt: 73…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02682026 CAS-Nummer: 51376-06-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...