6-Chinoxalincarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Quinoxaline-6-carboxylic acid
OR23160-1G
OR23160-5G
OR23160-25G
APOSOR23160-1GEA
21.9
EUR
APOSOR23160-1G
APOSOR23160-5G
APOSOR23160-25G
6-Chinoxalincarbonsäure
6-Quinoxalinecarboxylic acid
Formel:
C₉H₆N₂O₂ Molecular Weight: 174,16 g/mol Schmelzpunkt: 248…251 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01365834 CAS-Nummer: 6925-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...