3-Benzo[b]thiophensulfonylchlorid 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-Benzothiophene-3-sulphonyl Chloride, Benzothiophene-3-sulphonyl chloride
OR23173-1G
OR23173-250MG
APOSOR23173-1GEA
132
EUR
APOSOR23173-1G
APOSOR23173-250MG
3-Benzo[b]thiophensulfonylchlorid 95%
3-Benzo[b]thiophensulfonylchlorid
Formel:
C₈H₅ClO₂S₂ Molecular Weight: 232.71 g/mol Schmelzpunkt: 85…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02681897 CAS-Nummer: 18494-87-6 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...