4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-sulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3,4-Dihydro-4-methyl-2H-1,4-benzoxazine-7-sulphonyl chloride
OR23218-1G
OR23218-250MG
OR23218-100MG
OR23218-500MG
APOSOR23218-1GEA
1040
EUR
APOSOR23218-1G
APOSOR23218-250MG
APOSOR23218-100MG
APOSOR23218-500MG
4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-sulphonyl chloride
4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-sulphonyl chloride
Formel:
C₉H₁₀ClNO₃S Molecular Weight: 247,7 g/mol Schmelzpunkt: 44…45 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02681911 CAS-Nummer: 368869-93-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...