4-Methyl-2,3-dihydro-2H-1,4-benzoxazine-7-carbaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Methyl-3,4-dihydro-2H-benzo[b][1,4]oxazine-7-carbaldehyde, 3,4-Dihydro-4-methyl-2H-1,4-benzoxazine-7-carboxaldehyde

OR23219-1G OR23219-250MG
APOSOR23219-1GEA 0 EUR
APOSOR23219-1G APOSOR23219-250MG
4-Methyl-2,3-dihydro-2H-1,4-benzoxazine-7-carbaldehyde
4-Methyl-2,3-dihydro-2H-1,4-benzoxazine-7-carbaldehyde
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
Schmelzpunkt: 69…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02681912
CAS-Nummer: 141103-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR