5-(1,3-Oxazol-5-yl)thiophene-2-sulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR23225-1G
OR23225-250MG
APOSOR23225-1GEA
277
EUR
APOSOR23225-1G
APOSOR23225-250MG
5-(1,3-Oxazol-5-yl)thiophene-2-sulphonyl chloride
5-(1,3-Oxazol-5-yl)thiophene-2-sulphonyl chloride
Formel:
C₇H₄ClNO₃S₂ Molecular Weight: 249.7 g/mol Schmelzpunkt: 80…82 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02681961 CAS-Nummer: 321309-40-4 UN: 1759 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...