1H-Benzotriazol-5-carbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1H-Benzo[d][1,2,3]triazole-5-carboxylic acid, 5-Carboxybenzotriazole
OR23258-100G
OR23258-25G
OR23258-5G
APOSOR23258-100GEA
125
EUR
APOSOR23258-100G
APOSOR23258-25G
APOSOR23258-5G
1H-Benzotriazol-5-carbonsäure
1H-Benzotriazol-5-carbonsäure
Formel:
C₇H₅N₃O₂ Molecular Weight: 163.14 g/mol Schmelzpunkt: 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012318 CAS-Nummer: 23814-12-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...