Desoxy-Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]desoxycholamid)
Lieferant: Apollo Scientific
Achtung
Synonyme: Deoxy-bigchap
OR2325T-1G OR2325T-250MG
APOSOR2325T-1GEA 328 EUR
APOSOR2325T-1G APOSOR2325T-250MG
Desoxy-Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]desoxycholamid)
Desoxy-Big CHAP (N,N-Bis[3-(D-gluconamido)propyl]desoxycholamid)
A nonionic detergent with properties comparable to CHAPS & CHAPSO but with reduced electrostatic interactions.
Formel: C₄₂H₇₅N₃O₁₅
Molecular Weight: 862.05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00161482
CAS-Nummer: 86303-23-3