2-Phenyl-4-thiazolcarbonsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Phenylthiazole-4-carboxylic acid

OR23282-1G OR23282-5G
APOSOR23282-1GEA 30.6 EUR
APOSOR23282-1G APOSOR23282-5G
2-Phenyl-4-thiazolcarbonsäure
2-Phenyl-4-thiazolecarboxylic acid
Formel: C₁₀H₇NO₂S
Molecular Weight: 205,23 g/mol
Schmelzpunkt: 173…177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00141954
CAS-Nummer: 7113-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR