4-(1H-Imidazol-1-yl)benzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(1-Imidazolyl)benzaldehyde, 1-(4-Formylphenyl)imidazole, 4-(1H-Imidazol-1-yl)benzaldehyde
OR23299-1G
OR23299-5G
OR23299-10G
APOSOR23299-1GEA
21.9
EUR
APOSOR23299-1G
APOSOR23299-5G
APOSOR23299-10G
4-(1H-Imidazol-1-yl)benzaldehyd
4-Imidazol-1-ylbenzaldehyde
Formel:
C₁₀H₈N₂O Molecular Weight: 172,1833 g/mol Schmelzpunkt: 153…155 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00209977 CAS-Nummer: 10040-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...