4-(1H-Imidazol-1-yl)benzonitril

Lieferant: Apollo Scientific

Synonyme: 4-(1-Imidazolyl)benzonitrile, 1-(4-Cyanophenyl)imidazole, 4-(1H-imidazol-1-yl)benzonitrile

OR23301-5G
APOSOR23301-5GEA 75.4 EUR
APOSOR23301-5G
4-(1H-Imidazol-1-yl)benzonitril
4-(1H-Imidazol-1-yl)benzonitrile
Formel: C₁₀H₇N₃
Molecular Weight: 169,1800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00127147
CAS-Nummer: 25372-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR