4-(1H-1,2,4-Triazol-1-yl)benzoesäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-[1,2,4]Triazol-1-yl-benzoic acid

OR23304-1G OR23304-250MG
APOSOR23304-1GEA 59.6 EUR
APOSOR23304-1G APOSOR23304-250MG
4-(1H-1,2,4-Triazol-1-yl)benzoesäure
4-(1H-1,2,4-Triazol-1-yl)benzoesäure
Formel: C₉H₇N₃O₂
Molecular Weight: 189.1708 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02682060
CAS-Nummer: 162848-16-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR