4-(1H-1,2,4-Triazol-1-yl)benzaldehyd

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Formylphenyl)-1,2,4-triazole, 4-(1,2,4-Triazol-1-yl)benzaldehyde

OR23305-1G OR23305-5G
APOSOR23305-1GEA 21.9 EUR
APOSOR23305-1G APOSOR23305-5G
4-(1H-1,2,4-Triazol-1-yl)benzaldehyd
4-(1H-1,2,4-Triazol-1-yl)benzaldehyd
Formel: C₉H₇N₃O
Molecular Weight: 173.1714 g/mol
Schmelzpunkt: 149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02681969
CAS-Nummer: 27996-86-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR