(4-(1H-1,2,4-Triazol-1-yl)phenyl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(1,2,4-Triazol-1-yl)benzyl Alcohol
OR23307-1G
OR23307-5G
APOSOR23307-1GEA
42.2
EUR
APOSOR23307-1G
APOSOR23307-5G
(4-(1H-1,2,4-Triazol-1-yl)phenyl)methanol
(4-(1H-1,2,4-Triazol-1-yl)phenyl)methanol
Formel:
C₉H₉N₃O Molecular Weight: 175,1873 g/mol Schmelzpunkt: 124…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02682061 CAS-Nummer: 143426-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...