3-(2-Methyl-1,3-thiazol-4-yl)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

OR23313-1G OR23313-250MG OR23313-5G
APOSOR23313-1GEA 148 EUR
APOSOR23313-1G APOSOR23313-250MG APOSOR23313-5G
3-(2-Methyl-1,3-thiazol-4-yl)benzoic acid 95%
3-(2-Methyl-1,3-thiazol-4-yl)benzoic acid
Formel: C₁₁H₉NO₂S
Schmelzpunkt: 200…201 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02682065
CAS-Nummer: 28077-41-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR