6-Phenoxypyridin-3-amine
Lieferant: Apollo Scientific
|
Achtung
|
OR23323-1G
OR23323-25G
OR23323-5G
APOSOR23323-1GEA
21.9
EUR
APOSOR23323-1G
APOSOR23323-25G
APOSOR23323-5G
6-Phenoxypyridin-3-amine
6-Phenoxypyridin-3-amine
Formel:
C₁₁H₁₀N₂O Molecular Weight: 186.22 g/mol Schmelzpunkt: 71…72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01692449 CAS-Nummer: 25194-67-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...