(1-Pyrimidin-2-ylpiperidin-4-yl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
OR23342-1G
APOSOR23342-1GEA
154
EUR
APOSOR23342-1G
(1-Pyrimidin-2-ylpiperidin-4-yl)methanol
(1-Pyrimidin-2-ylpiperidin-4-yl)methanol
Formel:
C₁₀H₁₅N₃O Molecular Weight: 193,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03086127 CAS-Nummer: 111247-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...