3-Pyrrol-1-ylbenzoic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(1H-Pyrrol-1-yl)benzoic acid

OR23348-250MG
APOSOR23348-250MGEA 21.9 EUR
APOSOR23348-250MG
3-Pyrrol-1-ylbenzoic acid 97%
3-Pyrrol-1-ylbenzoic acid
Formel: C₁₁H₉NO₂
Molecular Weight: 187,2 g/mol
Schmelzpunkt: 175…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02656610
CAS-Nummer: 61471-45-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR