1,3-dimethyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,3-Dimethyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid
OR23361-1G
OR23361-5G
OR23361-100MG
APOSOR23361-1GEA
68.3
EUR
APOSOR23361-1G
APOSOR23361-5G
APOSOR23361-100MG
1,3-dimethyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid 95%
1,3-dimethyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid
Formel:
C₈H₈N₂O₂S Molecular Weight: 196.224 g/mol Schmelzpunkt: 260 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00126746 CAS-Nummer: 25252-46-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...