2,3-Dihydro-1-benzofuran-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydrobenzofuran-2-carboxylic acid

OR23376-1G OR23376-5G
APOSOR23376-1GEA 87 EUR
APOSOR23376-1G APOSOR23376-5G
2,3-Dihydro-1-benzofuran-2-carboxylic acid
2,3-Dihydro-1-benzofuran-2-carboxylic acid
Formel: C₉H₈O₃
Schmelzpunkt: 116…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03086168
CAS-Nummer: 1914-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR