5-Phenylnicotinic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR23382-1G
OR23382-250MG
APOSOR23382-1GEA
0
EUR
APOSOR23382-1G
APOSOR23382-250MG
5-Phenylnicotinic acid
5-Phenylnicotinic acid
Formel:
C₁₂H₉NO₂ Schmelzpunkt: 260 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03086176 CAS-Nummer: 10177-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...