[2-(4-Morpholinyl)phenyl]methanamine 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-(Morpholin-4-yl)benzylamine
OR23385-1G
OR23385-5G
APOSOR23385-1GEA
137
EUR
APOSOR23385-1G
APOSOR23385-5G
[2-(4-Morpholinyl)phenyl]methanamine 97%
[2-(4-Morpholinyl)phenyl]methanamine
Formel:
C₁₁H₁₆N₂O Molecular Weight: 192,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03086183 CAS-Nummer: 204078-48-8 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...