1-Phenyl-1H-pyrazol-5-carbonsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR23386-1G
OR23386-250MG
APOSOR23386-1GEA
109
EUR
APOSOR23386-1G
APOSOR23386-250MG
1-Phenyl-1H-pyrazol-5-carbonsäure 97%
1-Phenyl-1H-pyrazole-5-carboxylic acid
Formel:
C₁₀H₈N₂O₂ Molecular Weight: 188,180 g/mol Schmelzpunkt: 182…186 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00463580 CAS-Nummer: 1133-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...