2-Chlor-3-iodpyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR2342-100G
OR2342-25G
OR2342-5G
APOSOR2342-100GEA
120
EUR
APOSOR2342-100G
APOSOR2342-25G
APOSOR2342-5G
2-Chlor-3-iodpyridin
2-Chlor-3-iodpyridin
Formel:
C₅H₃ClIN Molecular Weight: 239,44 g/mol Schmelzpunkt: 93…97 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00661298 CAS-Nummer: 78607-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...